accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSOOHFOHOF

CAS RN 147027-64-3

IUPAC Name: 2-[(2-carboxy-3-fluorophenyl)disulfanyl]-6-fluorobenzoic acid
Synonyms: 2-[(2-carboxy-3-fluorophenyl)disulfanyl]-6-fluorobenzoic acid
Molecular Weight (g/mol): 342.33
Molecular Formula: C14H8F2O4S2
InChi Key: QEONSZINEMHFPH-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC=C1SSC1=CC=CC(F)=C1C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results