accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONH2N

CAS RN 147081-44-5

IUPAC Name: tert-butyl 3-aminopyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-aminopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: CMIBWIAICVBURI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(N)C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)