accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14716-89-3

IUPAC Name: (3-methyl-1,2-oxazol-5-yl)methanol
Synonyms: (3-methyl-1,2-oxazol-5-yl)methanol
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: OFSDWHRZVRCPBR-UHFFFAOYSA-N
SMILES: CC1=NOC(CO)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results