accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1472-93-1

IUPAC Name: 1,18-dimethyl octadecanedioate
Synonyms: 1,18-dimethyl octadecanedioate
Molecular Weight (g/mol): 342.52
Molecular Formula: C20H38O4
InChi Key: ZOXMHKCFDXHCJX-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCCCCCCCCCC(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results