accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 147266-92-0

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 231.25
Molecular Formula: C10H17NO5
InChi Key: BENKAPCDIOILGV-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results