accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 147290-10-6

OHOOOHNH2N(S)CH2

CAS RN 147290-10-6

IUPAC Name: (2S)-2-amino-5-{[(prop-2-en-1-yloxy)carbonyl]amino}pentanoic acid
Synonyms: (2S)-2-amino-5-{[(prop-2-en-1-yloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 216.24
Molecular Formula: C9H16N2O4
InChi Key: PIBIUFMNGVYLNI-ZETCQYMHSA-N
SMILES: N[C@@H](CCCNC(=O)OCC=C)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)