accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNNH2

CAS RN 147291-66-5

IUPAC Name: tert-butyl N-[(3-aminophenyl)methyl]carbamate
Synonyms: tert-butyl N-[(3-aminophenyl)methyl]carbamate
Molecular Weight (g/mol): 222.29
Molecular Formula: C12H18N2O2
InChi Key: LSOZALWRNWQPLK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC(N)=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)