accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14753-05-0

IUPAC Name: 1-chloro-2-methylpentane
Synonyms: 1-chloro-2-methylpentane
Molecular Weight (g/mol): 120.62
Molecular Formula: C6H13Cl
InChi Key: SLBTUZZYJLBZQV-UHFFFAOYNA-N
SMILES: CCCC(C)CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results