accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1476-23-9

IUPAC Name: 3-isocyanatoprop-1-ene
Synonyms: 1-propene, 3-isocyanato-
Molecular Weight (g/mol): 83.09
Molecular Formula: C4H5NO
InChi Key: HXBPYFMVGFDZFT-UHFFFAOYSA-N
SMILES: C=CCN=C=O
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)