accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONBr

CAS RN 147621-26-9

IUPAC Name: tert-butyl 6-bromo-1H-indole-1-carboxylate
Synonyms: tert-butyl 6-bromoindole-1-carboxylate
Molecular Weight (g/mol): 296.16
Molecular Formula: C13H14BrNO2
InChi Key: OUOWFTDOMYJYKB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC2=CC=C(Br)C=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)