accessibility menu, dialog, popup

UserName

Viewing profile as user:
GdOOOOOOCCCCH3H3CH3CCH3CH3CH3CH3H3CCH3H3CH3CCH3CH3CH3H3CH3CCH3H3CHHH

CAS RN 14768-15-1

IUPAC Name: gadolinium(3+) tris(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Synonyms: gadolinium(3+) tris(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular Weight (g/mol): 707.06
Molecular Formula: C33H57GdO6
InChi Key: UIVVGOMXFUOLAN-UHFFFAOYSA-N
SMILES: [Gd+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results