accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOOH(E/Z)

CAS RN 147700-58-1

IUPAC Name: 3-(3,5-difluorophenyl)prop-2-enoic acid
Synonyms: 3-(3,5-difluorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 184.14
Molecular Formula: C9H6F2O2
InChi Key: MBAWRXICVNIUGY-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(F)=CC(F)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results