accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 147923-08-8

IUPAC Name: (2S)-3-{[1,1'-biphenyl]-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2S)-3-{[1,1'-biphenyl]-4-yl}-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 341.41
Molecular Formula: C20H23NO4
InChi Key: NBVVKAUSAGHTSU-KRWDZBQOSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results