accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14794-31-1

IUPAC Name: ethyl 4-chloro-4-oxobutanoate
Synonyms: ethyl 4-chloro-4-oxobutanoate
Molecular Weight (g/mol): 164.59
Molecular Formula: C6H9ClO3
InChi Key: IXZFDJXHLQQSGQ-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(Cl)=O
Boiling Point 
  • (2)
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (3)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (4)