accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 147951-36-8

IUPAC Name: 3',5'-bis[4-(diphenylamino)phenyl]-N,N-diphenyl-[1,1'-biphenyl]-4-amine
Synonyms: 3',5'-bis[4-(diphenylamino)phenyl]-N,N-diphenyl-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 808.04
Molecular Formula: C60H45N3
InChi Key: XVMUGTFNHXHZIP-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results