accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 148-69-6

IUPAC Name: 3-[ethyl(3-methylphenyl)amino]propanenitrile
Synonyms: 3-[ethyl(3-methylphenyl)amino]propanenitrile
Molecular Weight (g/mol): 188.27
Molecular Formula: C12H16N2
InChi Key: NPCCPMHHNIOSHL-UHFFFAOYSA-N
SMILES: CCN(CCC#N)C1=CC=CC(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results