accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 148-72-1

IUPAC Name: (3S)-3-ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-one; nitric acid
Synonyms: (3S)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one; nitric acid
Molecular Weight (g/mol): 271.27
Molecular Formula: C11H17N3O5
InChi Key: PRZXEPJJHQYOGF-LQRGNCEWSA-N
SMILES: O[N+]([O-])=O.CC[C@H]1C(CC2=CN=CN2C)COC1=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results