accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFNOO

CAS RN 1480-87-1

IUPAC Name: 2-fluoro-3-nitropyridine
Synonyms: 2-fluoro-3-nitropyridine
Molecular Weight (g/mol): 142.09
Molecular Formula: C5H3FN2O2
InChi Key: QDKIYDGHCFZBGC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)N=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)