accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14804-39-8

IUPAC Name: 2-methoxy-1,3-dimethyl-5-nitrobenzene
Synonyms: 2-methoxy-1,3-dimethyl-5-nitrobenzene
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: HSDNHFOJTRMGER-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(C=C1C)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results