accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 14811-73-5

IUPAC Name: methyl 10-oxodecanoate
Synonyms: methyl 10-oxodecanoate
Molecular Weight (g/mol): 200.28
Molecular Formula: C11H20O3
InChi Key: LVPNLDGNIRVDRF-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results