accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OHHC

CAS RN 1482-15-1

IUPAC Name: 3,4-dimethylpent-1-yn-3-ol
Synonyms: 1-pentyn-3-ol, 3,4-dimethyl-
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: DZNLEQBXXLGELU-UHFFFAOYNA-N
SMILES: CC(C)C(C)(O)C#C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results