accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSBrBr

CAS RN 148256-63-7

IUPAC Name: 2,5-dibromo-3-dodecylthiophene
Synonyms: 2,5-dibromo-3-dodecylthiophene
Molecular Weight (g/mol): 410.25
Molecular Formula: C16H26Br2S
InChi Key: ZIZGNWAMVWNAJT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=C(Br)SC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results