accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3SH

CAS RN 1483-27-8

IUPAC Name: 2,5-dimethoxybenzene-1-thiol
Synonyms: 2,5-dimethoxybenzenethiol
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: SESUUAOAUZDHHP-UHFFFAOYSA-N
SMILES: COC1=CC(S)=C(OC)C=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results