accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFFN

CAS RN 1483-54-1

IUPAC Name: 2-amino-3-(trifluoromethyl)benzonitrile
Synonyms: 2-amino-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 186.14
Molecular Formula: C8H5F3N2
InChi Key: UHOSMRSMWHLHJO-UHFFFAOYSA-N
SMILES: NC1=C(C=CC=C1C(F)(F)F)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results