accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1484-26-0

IUPAC Name: 3-(benzyloxy)aniline
Synonyms: 3-(benzyloxy)aniline
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: IGPFOKFDBICQMC-UHFFFAOYSA-N
SMILES: NC1=CC=CC(OCC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)