accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1484-85-1

IUPAC Name: hydrogen 2-(2H-1,3-benzodioxol-5-yl)ethan-1-amine chloride
Synonyms: hydrogen 2-(2H-1,3-benzodioxol-5-yl)ethanamine chloride
Molecular Weight (g/mol): 201.65
Molecular Formula: C9H12ClNO2
InChi Key: NDYXFQODWGEGNU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results