accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 148451-96-1

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
Synonyms: [3,5-bis(trifluoromethyl)phenyl]methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 472.39
Molecular Formula: C22H18F6N2O3
InChi Key: BYYQYXVAWXAYQC-IBGZPJMESA-N
SMILES: CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)