accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NH3CCH3OOCH3

CAS RN 148491-00-3

IUPAC Name: 2-methoxy-N,N-bis(propan-2-yl)benzamide
Synonyms: N,N-diisopropyl-2-methoxybenzamide
Molecular Weight (g/mol): 235.33
Molecular Formula: C14H21NO2
InChi Key: JIRXADQFESAWGU-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)N(C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results