accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOO(Z)

CAS RN 1485-00-3

IUPAC Name: 5-[(1Z)-2-nitroethenyl]-2H-1,3-benzodioxole
Synonyms: 5-[(1Z)-2-nitroethenyl]-2H-1,3-benzodioxole
Molecular Weight (g/mol): 193.16
Molecular Formula: C9H7NO4
InChi Key: KFLWBZPSJQPRDD-ARJAWSKDSA-N
SMILES: [O-][N+](=O)\C=C/C1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results