accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNCH3HOOH

CAS RN 148546-82-1

IUPAC Name: (4-methylpyridin-3-yl)boronic acid
Synonyms: 4-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: ASXFMIDIRZPCGK-UHFFFAOYSA-N
SMILES: CC1=C(C=NC=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results