CAS RN 1487-82-7
IUPAC Name:
2-[4-(dicyanomethylidene)-2,5-dimethylcyclohexa-2,5-dien-1-ylidene]propanedinitrile
Synonyms:
2-[4-(dicyanomethylidene)-2,5-dimethylcyclohexa-2,5-dien-1-ylidene]propanedinitrile
Molecular Weight (g/mol):
232.25
Molecular Formula:
C14H8N4
InChi Key:
DFJXWQJAMNCPII-UHFFFAOYSA-N
SMILES:
CC1=CC(=C(C#N)C#N)C(C)=CC1=C(C#N)C#N
1
–
1
of
1
results
1
2,5-Dimethyl-7,7,8,8-tetracyanoquinodimethane 98.0+%, TCI America™
CAS: 1487-82-7 Molecular Formula: C14H8N4 Molecular Weight (g/mol): 232.25 MDL Number: MFCD00130123 InChI Key: DFJXWQJAMNCPII-UHFFFAOYSA-N PubChem CID: 616047 IUPAC Name: 2-[4-(dicyanomethylidene)-2,5-dimethylcyclohexa-2,5-dien-1-ylidene]propanedinitrile SMILES: CC1=CC(=C(C#N)C#N)C(C)=CC1=C(C#N)C#N