accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 148705-09-3

IUPAC Name: N-{2-[(2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl)oxy]-5-hydroxy-6-(hydroxymethyl)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-3-yl}acetamide
Synonyms: N-{2-[(2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl)oxy]-5-hydroxy-6-(hydroxymethyl)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-3-yl}acetamide
Molecular Weight (g/mol): 828.73
Molecular Formula: C32H48N2O23
InChi Key: UUSZRTFVRDAQGP-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(OC2C(O)C(CO)OC(OC3C(O)C(O)C(OC4=CC=C(C=C4)[N+]([O-])=O)OC3CO)C2O)OC(CO)C(O)C1OC1OC(CO)C(O)C(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results