accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3MgBr

CAS RN 148706-30-3

IUPAC Name: bromo(4-methoxy-2-methylphenyl)magnesium
Synonyms: bromo(4-methoxy-2-methylphenyl)magnesium
Molecular Weight (g/mol): 225.37
Molecular Formula: C8H9BrMgO
InChi Key: JRXYWLSUMCREBA-UHFFFAOYSA-M
SMILES: COC1=CC=C([Mg]Br)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results