accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14872-21-0

IUPAC Name: dichloroplatinum; bis(pyridine)
Synonyms: platinum(II) chloride; bis(pyridine)
Molecular Weight (g/mol): 424.19
Molecular Formula: C10H10Cl2N2Pt
InChi Key: FCJXCLMZOWAKNQ-UHFFFAOYSA-L
SMILES: Cl[Pt]Cl.C1=CC=NC=C1.C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results