accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HSOHOH2NHCl

CAS RN 148766-37-4

IUPAC Name: 2-amino-2-methyl-3-sulfanylpropanoic acid hydrochloride
Synonyms: 2-amino-2-methyl-3-sulfanylpropanoic acid hydrochloride
Molecular Weight (g/mol): 171.64
Molecular Formula: C4H10ClNO2S
InChi Key: MAGCVRLGTQSVGF-UHFFFAOYNA-N
SMILES: Cl.CC(N)(CS)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results