accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSnH3CCH3S

CAS RN 148961-88-0

IUPAC Name: (1-benzothiophen-2-yl)tributylstannane
Synonyms: 1-benzothiophen-2-yltributylstannane
Molecular Weight (g/mol): 423.25
Molecular Formula: C20H32SSn
InChi Key: UJPVMOUQRNYNSA-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)