accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14899-31-1

IUPAC Name: 3-phosphonobenzoic acid
Synonyms: 3-phosphonobenzoic acid
Molecular Weight (g/mol): 202.1
Molecular Formula: C7H7O5P
InChi Key: ZKKXCRILZNBJJM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)P(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results