CAS RN 149-91-7
Gallic acid, 98%
CAS: 149-91-7 Molecular Formula: C7H6O5 Molecular Weight (g/mol): 170.12 MDL Number: MFCD00002510 InChI Key: LNTHITQWFMADLM-UHFFFAOYSA-N Synonym: gallic acid,gallate,benzoic acid, 3,4,5-trihydroxy,gallic acid, tech.,galop,pyrogallol-5-carboxylic acid,kyselina gallova,3,4,5-trihydroxybenzoate,ccris 5523,kyselina gallova czech PubChem CID: 370 ChEBI: CHEBI:30778 IUPAC Name: 3,4,5-trihydroxybenzoic acid SMILES: OC(=O)C1=CC(O)=C(O)C(O)=C1
Gallic Acid, MP Biomedicals
CAS: 149-91-7 Molecular Formula: C7H6O5 Molecular Weight (g/mol): 170.12 MDL Number: MFCD00002510 InChI Key: LNTHITQWFMADLM-UHFFFAOYSA-N Synonym: gallic acid,gallate,benzoic acid, 3,4,5-trihydroxy,gallic acid, tech.,galop,pyrogallol-5-carboxylic acid,kyselina gallova,3,4,5-trihydroxybenzoate,ccris 5523,kyselina gallova czech PubChem CID: 370 ChEBI: CHEBI:30778 IUPAC Name: 3,4,5-trihydroxybenzoic acid SMILES: OC(=O)C1=CC(O)=C(O)C(O)=C1
3,4,5-Trihydroxybenzoic acid, 98%
CAS: 149-91-7 Molecular Formula: C7H6O5 Molecular Weight (g/mol): 170.12 MDL Number: MFCD00002510 InChI Key: LNTHITQWFMADLM-UHFFFAOYSA-N Synonym: gallic acid,gallate,benzoic acid, 3,4,5-trihydroxy,gallic acid, tech.,galop,pyrogallol-5-carboxylic acid,kyselina gallova,3,4,5-trihydroxybenzoate,ccris 5523,kyselina gallova czech PubChem CID: 370 ChEBI: CHEBI:30778 IUPAC Name: 3,4,5-trihydroxybenzoic acid SMILES: OC(=O)C1=CC(O)=C(O)C(O)=C1
phyproof™ Gallic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Gallic Acid, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Gallic Acid, Monohydrate, ACS Grade, AquaPhoenix Scientific™
CAS: 149-91-7 Molecular Formula: C7H6O5 Molecular Weight (g/mol): 170.12 MDL Number: MFCD00002510 InChI Key: LNTHITQWFMADLM-UHFFFAOYSA-N IUPAC Name: 3,4,5-trihydroxybenzoic acid SMILES: OC(=O)C1=CC(O)=C(O)C(O)=C1