accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1490-25-1

IUPAC Name: methyl 4-chloro-4-oxobutanoate
Synonyms: methyl 4-chloro-4-oxobutanoate
Molecular Weight (g/mol): 150.56
Molecular Formula: C5H7ClO3
InChi Key: SRXOJMOGPYFZKC-UHFFFAOYSA-N
SMILES: COC(=O)CCC(Cl)=O
Boiling Point 
  • (3)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (1)
  • (6)
  • (2)
  • (3)
  • (2)
  • (1)