accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)N(R)O(R)(S)PNNOOOHOOHNH2HONa

CAS RN 149022-20-8

IUPAC Name: sodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
Synonyms: NaAmp
Molecular Weight (g/mol): 369.21
Molecular Formula: C10H13N5NaO7P
InChi Key: FYWLYWMEGHCZAX-GWKNMROSNA-M
SMILES: [Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)