accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 14906-59-3

IUPAC Name: 4-cyanopyridin-1-ium-1-olate
Synonyms: 4-cyanopyridine oxide
Molecular Weight (g/mol): 120.11
Molecular Formula: C6H4N2O
InChi Key: QNCSFBSIWVBTHE-UHFFFAOYSA-N
SMILES: [O-][N+]1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)
  • (2)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)