CAS RN 14906-64-0
CAS RN 14906-64-0
IUPAC Name:
3-cyano-1,4-dihydro-1λ²-pyridine-1,1-diium-4-id-1-olate
Synonyms:
3-cyano-1,4-dihydro-1λ²-pyridine-1,1-diium-4-id-1-olate
Molecular Weight (g/mol):
121.12
Molecular Formula:
C6H5N2O
InChi Key:
AABSWFBNBQBPEQ-UHFFFAOYNA-N
SMILES:
[O-][NH++]1C=C[CH-]C(=C1)C#N
1
–
1
of
1
results
1
Thermo Scientific Chemicals 3-Cyanopyridine 1-oxide, 97%
CAS: 14906-64-0 Molecular Formula: C6H5N2O Molecular Weight (g/mol): 121.12 MDL Number: MFCD00129037 InChI Key: AABSWFBNBQBPEQ-UHFFFAOYNA-N Synonym: nicotinonitrile-1-oxide,3-cyanopyridine n-oxide,3-cyanopyridine-n-oxide,nicotinonitrile 1-oxide,3-cyanopyridine 1-oxide,3-cyanopyridin-1-ium-1-olate,3-pyridinecarbonitrile, 1-oxide,acmc-1c6hr,3-pyridinecarbonitrile,1-oxide,woovsqcalyyudo-uhfffaoysa PubChem CID: 84686 SMILES: [O-][NH++]1C=C[CH-]C(=C1)C#N