accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOOOLa

CAS RN 14913-14-5

IUPAC Name: lanthanum(3+) phosphate
Synonyms: lanthanum phosphate
Molecular Weight (g/mol): 233.88
Molecular Formula: LaO4P
InChi Key: LQFNMFDUAPEJRY-UHFFFAOYSA-K
SMILES: [La+3].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results