accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OHoClClCl

CAS RN 14914-84-2

IUPAC Name: trichloroholmium hexahydrate
Synonyms: trichloroholmium hexahydrate
Molecular Weight (g/mol): 379.37
Molecular Formula: Cl3H12HoO6
InChi Key: BUQFCXABQYNXLP-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.Cl[Ho](Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)