accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFNNNNNFFFFNNNFFFFFFFFCu

CAS RN 14916-87-1

IUPAC Name: copper(2+) hexadecafluoro-2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Synonyms: copper(2+) hexadecafluoro-2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Molecular Weight (g/mol): 863.93
Molecular Formula: C32CuF16N8
InChi Key: FJAOBQORBYMRNO-UHFFFAOYSA-N
SMILES: [Cu++].FC1=C2C3=NC4=NC(=NC5=C6C(=C([N-]5)N=C5N=C(N=C([N-]3)C2=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C51)C(F)=C(F)C(F)=C6F)C1=C(F)C(F)=C(F)C(F)=C41
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results