CAS RN 14918-69-5
CAS RN 14918-69-5
IUPAC Name:
6,7-dichloro-5,8-dihydroxy-1,4-dihydronaphthalene-1,4-dione
Synonyms:
6,7-dichloro-5,8-dihydroxynaphthalene-1,4-dione
Molecular Weight (g/mol):
259.04
Molecular Formula:
C10H4Cl2O4
InChi Key:
KFAVXUQFBGHURZ-UHFFFAOYSA-N
SMILES:
OC1=C(Cl)C(Cl)=C(O)C2=C1C(=O)C=CC2=O
1
–
1
of
1
results
1
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone 97.0+%, TCI America™
CAS: 14918-69-5 Molecular Formula: C10H4Cl2O4 Molecular Weight (g/mol): 259.038 MDL Number: MFCD00075261 InChI Key: KFAVXUQFBGHURZ-UHFFFAOYSA-N PubChem CID: 4407 IUPAC Name: 6,7-dichloro-5,8-dihydroxynaphthalene-1,4-dione SMILES: C1=CC(=O)C2=C(C1=O)C(=C(C(=C2O)Cl)Cl)O