accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14919-01-8

IUPAC Name: (3E)-oct-3-ene
Synonyms: (3E)-oct-3-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: YCTDZYMMFQCTEO-FNORWQNLSA-N
SMILES: CCCC\C=C\CC
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results