accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14920-87-7

IUPAC Name: methyl 2,6-dichlorobenzoate
Synonyms: methyl 2,6-dichlorobenzoate
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: HEYGSGDIWJDORA-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results