accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3C

CAS RN 14920-89-9

IUPAC Name: 2,3-dimethylfuran
Synonyms: furan, dimethyl-
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: FJSKXQVRKZTKSI-UHFFFAOYSA-N
SMILES: CC1=C(C)C=CO1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)