accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14933-08-5

IUPAC Name: 3-(dodecyldimethylazaniumyl)propane-1-sulfonate
Synonyms: lauryl sultaine
Molecular Weight (g/mol): 335.55
Molecular Formula: C17H37NO3S
InChi Key: IZWSFJTYBVKZNK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
Quantity 
  • (1)
  • (4)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)